Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000157.3 | NP_000148.2 | 8 | 1181 | |
NM_001005741.2 | NP_001005741.1 | 9 | 1440 | ||
NM_001005742.2 | NP_001005742.1 | 9 | 1421 | ||
NM_001171811.1 | NP_001165282.1 | 7 | 1257 | ||
NM_001171812.1 | NP_001165283.1 | 7 | 1031 | ||
GenBank mRNA | AK291911.1 | - | 9 | 1419 | |
AK298900.1 | - | 7 | 1031 | ||
AK300186.1 | - | 7 | 1101 | ||
AK300829.1 | - | 7 | 1094 | ||
AK300876.1 | - | 9 | 1421 | ||
AK301374.1 | - | 6 | 790 | ||
AK301879.1 | - | 7 | 760 | ||
AK302000.1 | - | 7 | 1006 | ||
AK312502.1 | - | 9 | 1237 | ||
BC003356.1 | - | 8 | 1120 | 2899915 | |
BX648487.1 | - | 9 | 1249 | ||
D13286.1 | - | 8 | 1120 | ||
K02920.1 | - | 7 | 1000 | ||
KJ690771.1 | - | 8 | 1064 | ||
KJ690773.1 | - | 8 | 1064 | ||
M16328.1 | - | 8 | 1450 | ||
M19285.1 | - | 8 | 1137 |