Gene Transcripts

Gene Symbol
SIAH3
Entrez Gene ID
283514
Gene Name
siah E3 ubiquitin protein ligase family member 3
Gene Aliases
Chromosome Location
Chr. 13: 45780270 - 45851711 on Build GRCh38
UniGene ID
Hs.368483
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198849.2 NP_942146.2 2 747
GenBank mRNA BC041372.2 - 2 747

View Full Product Details