Gene Transcripts
- Gene Symbol
- ADGRE2
- Entrez Gene ID
- 30817
- Gene Name
- adhesion G protein-coupled receptor E2
- Gene Aliases
- CD312, EMR2, VBU
- Chromosome Location
- Chr. 19: 14732697 - 14778541 on Build GRCh38
- UniGene ID
- Hs.531619
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271052.1 |
NP_001257981.1
|
16, 17 |
2182 |
|
NM_013447.3 |
NP_038475.2
|
18, 19 |
2641 |
|
XM_011527948.2 |
XP_011526250.1
|
17, 18 |
2270 |
|
XM_011527949.2 |
XP_011526251.1
|
17, 18 |
2237 |
|
XM_011527951.2 |
XP_011526253.1
|
16, 17 |
2123 |
|
XM_011527952.2 |
XP_011526254.1
|
16, 17 |
2096 |
|
XM_011527953.2 |
XP_011526255.1
|
15, 16 |
2077 |
|
XM_011527954.2 |
XP_011526256.1
|
14, 15 |
2808 |
|
XM_011527955.2 |
XP_011526257.1
|
12, 13 |
1687 |
|
XM_017026726.1 |
XP_016882215.1
|
16, 17 |
2239 |
|
XM_017026727.1 |
XP_016882216.1
|
15, 16 |
1916 |
|
GenBank mRNA |
AF114491.1 |
- |
17, 18 |
2254 |
|
AK024426.1 |
- |
8, 9 |
1010 |
|
AK024454.1 |
- |
3, 4 |
223 |
|
AK222891.1 |
- |
17, 18 |
2239 |
|
AK291776.1 |
- |
17, 18 |
2387 |
|
AK298690.1 |
- |
12, 13 |
1492 |
|
AK298700.1 |
- |
16, 17 |
2098 |
|
BC082991.1 |
- |
17, 18 |
2198 |
|
BC127004.1 |
- |
18, 19 |
2637 |
40108332 |
BC127005.1 |
- |
18, 19 |
2637 |
40108333 |
CR936819.1 |
- |
12, 13 |
1618 |
|
View Full Product Details