Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000601.5 | NP_000592.3 | 2 | 439 | |
NM_001010931.2 | NP_001010931.1 | 2 | 439 | ||
NM_001010932.2 | NP_001010932.1 | 2 | 439 | ||
NM_001010933.2 | NP_001010933.1 | 2 | 439 | ||
NM_001010934.2 | NP_001010934.1 | 2 | 439 | ||
XM_006715956.2 | XP_006716019.1 | 3 | 402 | ||
XM_011516115.2 | XP_011514417.1 | 3 | 402 | ||
XM_017012097.1 | XP_016867586.1 | 3 | 402 | ||
XM_017012098.1 | XP_016867587.1 | 3 | 402 | ||
GenBank mRNA | AB208900.1 | - | 2 | 346 | |
AK291674.1 | - | 2 | 374 | ||
AK313694.1 | - | 2 | 289 | ||
BC022308.1 | - | 2 | 288 | ||
BC063485.1 | - | 2 | 297 | ||
BC105797.1 | - | 2 | 379 | ||
BC130284.1 | - | 2 | 287 | 40146315 | |
BC130286.1 | - | 2 | 287 | 40146317 | |
BU533235.1 | - | 378 | |||
BX495370.1 | - | 2 | 440 | ||
FJ830861.1 | - | 2 | 212 | ||
FJ830862.1 | - | 2 | 212 | ||
L02931.1 | - | 2 | 272 | ||
M29145.1 | - | 2 | 313 | ||
M55379.1 | - | 2 | 212 | ||
M60718.1 | - | 2 | 284 | ||
M73239.1 | - | 2 | 277 | ||
M73240.1 | - | 2 | 277 | ||
M77227.1 | - | 2 | 294 | ||
U46010.1 | - | 2 | 266 | ||
X16323.1 | - | 2 | 346 | ||
X57574.1 | - | 2 | 289 |