Gene Transcripts

Gene Symbol
VN1R4
Entrez Gene ID
317703
Gene Name
vomeronasal 1 receptor 4
Gene Aliases
V1RL4
Chromosome Location
Chr. 19: 53264418 - 53270939 on Build GRCh38
UniGene ID
Hs.553685
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173857.2 NP_776256.2 1 539
XM_011526885.1 XP_011525187.1 2 691
XM_017026728.1 XP_016882217.1 3 439
XM_017026729.1 XP_016882218.1 3 457
XM_017026730.1 XP_016882219.1 3 398
GenBank mRNA BC104935.1 - 1 593
BC104939.1 - 1 597 8143942

View Full Product Details