Gene Transcripts

Gene Symbol
LRRC30
Entrez Gene ID
339291
Gene Name
leucine rich repeat containing 30
Gene Aliases
Chromosome Location
Chr. 18: 7231139 - 7232044 on Build GRCh38
UniGene ID
Hs.647588
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105581.1 NP_001099051.1 1 584

View Full Product Details