Gene Transcripts

Gene Symbol
MAGEB5
Entrez Gene ID
347541
Gene Name
MAGE family member B5
Gene Aliases
CT3.3, MAGE-B5
Chromosome Location
Chr. X: 26216169 - 26218270 on Build GRCh38
UniGene ID
Hs.224079
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271752.1 NP_001258681.1 2 351
GenBank mRNA CR747535.1 - 2 351

View Full Product Details