Gene Transcripts

Gene Symbol
C8orf86
Entrez Gene ID
389649
Gene Name
chromosome 8 open reading frame 86
Gene Aliases
Chromosome Location
Chr. 8: 38510834 - 38528662 on Build GRCh38
UniGene ID
Hs.546586
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207412.2 NP_997295.1 1, 2 330
GenBank mRNA AK125570.1 - 1, 2 330
BC137511.1 - 1, 2 325 9021511
BC137512.1 - 1, 2 325 9021512

View Full Product Details