Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002439.4 | NP_002430.3 | 6 | 1011 | |
GenBank mRNA | AK289856.1 | - | 7 | 980 | |
BC004177.2 | - | 7 | 952 | ||
BC011817.2 | - | 7 | 952 | ||
BC017273.2 | - | 6 | 934 | ||
BC098436.1 | - | 7 | 952 | ||
BC130434.1 | - | 6 | 1011 | 40146465 | |
BC130436.1 | - | 6 | 1011 | 40146467 | |
J04810.1 | - | 6 | 992 | ||
U61981.1 | - | 7 | 920 |