Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006166.3 | NP_006157.1 | 5 | 513 | |
XM_005268908.4 | XP_005268965.1 | 6 | 577 | ||
XM_005268909.4 | XP_005268966.1 | 6 | 560 | ||
XM_005268910.4 | XP_005268967.1 | 6 | 1232 | ||
GenBank mRNA | AK291934.1 | - | 5 | 445 | |
BC005316.1 | - | 5 | 462 | 3680937 | |
BC005317.1 | - | 5 | 468 | 3997299 | |
BC007035.1 | - | 5 | 440 | 3996244 | |
L06145.1 | - | 3 | 312 | ||
X59710.1 | - | 5 | 417 |