Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016216.3 | NP_057300.2 | 7, 8 | 1103 | |
GenBank mRNA | AF180919.2 | - | 7, 8 | 1057 | |
AK000116.1 | - | 2, 3 | 580 | ||
AK094147.1 | - | 8, 9 | 988 | ||
BC009472.1 | - | 7, 8 | 1031 | 3503627 |