Gene Transcripts

Gene Symbol
HSD17B11
Entrez Gene ID
51170
Gene Name
hydroxysteroid 17-beta dehydrogenase 11
Gene Aliases
17-BETA-HSD11, 17-BETA-HSDXI, 17BHSD11, DHRS8, PAN1B, RETSDR2, SDR16C2
Chromosome Location
Chr. 4: 87336522 - 87391303 on Build GRCh38
UniGene ID
Hs.594923
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016245.4 NP_057329.3 7 1098
GenBank mRNA AF126780.1 - 7 1053
AF273056.1 - 7 951
AK075348.1 - 7 982
AY358553.1 - 7 965
BC008650.1 - 7 988 3854750
BC014327.1 - 7 964 3961721
BC016367.1 - 7 995 4133318
BC021673.1 - 7 969 3827935
BC036001.1 - 7 989 4715042
CA775960.1 - 1 698

View Full Product Details