Gene Transcripts
- Gene Symbol
- SNX9
- Entrez Gene ID
- 51429
- Gene Name
- sorting nexin 9
- Gene Aliases
- SDP1, SH3PX1, SH3PXD3A, WISP
- Chromosome Location
- Chr. 6: 157716046 - 157945077 on Build GRCh38
- UniGene ID
- Hs.191213
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016224.4 |
NP_057308.1
|
13, 14 |
1627 |
|
XM_005267015.2 |
XP_005267072.1
|
15, 16 |
1705 |
|
XM_011535886.2 |
XP_011534188.1
|
10, 11 |
1166 |
|
GenBank mRNA |
AB209736.1 |
- |
9, 10 |
958 |
|
AF076957.1 |
- |
8, 9 |
847 |
|
AF121859.1 |
- |
13, 14 |
1387 |
|
AF131214.1 |
- |
13, 14 |
1406 |
|
AF172847.1 |
- |
13, 14 |
1507 |
|
AK026637.1 |
- |
10, 11 |
1172 |
|
AK127345.1 |
- |
10, 11 |
1482 |
|
AK314360.1 |
- |
13, 14 |
1461 |
|
BC001084.2 |
- |
4, 5 |
428 |
|
BC005022.1 |
- |
13, 14 |
1534 |
3832234 |
View Full Product Details