Gene Transcripts
- Gene Symbol
- TMCO3
- Entrez Gene ID
- 55002
- Gene Name
- transmembrane and coiled-coil domains 3
- Gene Aliases
- C13orf11
- Chromosome Location
- Chr. 13: 113490993 - 113550229 on Build GRCh38
- UniGene ID
- Hs.317593
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017905.4 |
NP_060375.4
|
4, 5 |
1173 |
|
XM_006719969.1 |
XP_006720032.1
|
4, 5 |
954 |
|
XM_011537498.2 |
XP_011535800.1
|
4, 5 |
1297 |
|
XM_011537501.2 |
XP_011535803.1
|
4, 5 |
1173 |
|
XM_011537502.2 |
XP_011535804.1
|
4, 5 |
1173 |
|
XM_011537504.2 |
XP_011535806.1
|
4, 5 |
1173 |
|
XM_011537506.2 |
XP_011535808.1
|
4, 5 |
1173 |
|
XM_011537507.2 |
XP_011535809.1
|
4, 5 |
1173 |
|
XM_011537509.2 |
XP_011535811.1
|
4, 5 |
1173 |
|
XM_017020629.1 |
XP_016876118.1
|
4, 5 |
1173 |
|
XM_017020630.1 |
XP_016876119.1
|
4, 5 |
1173 |
|
XM_017020633.1 |
XP_016876122.1
|
4, 5 |
1173 |
|
XM_017020634.1 |
XP_016876123.1
|
4, 5 |
1173 |
|
XM_017020636.1 |
XP_016876125.1
|
4, 5 |
1173 |
|
GenBank mRNA |
AF209505.1 |
- |
3, 4 |
387 |
|
AK000630.1 |
- |
4, 5 |
1070 |
|
AK074415.1 |
- |
1, 2 |
199 |
|
AY358768.1 |
- |
4, 5 |
951 |
|
BC012564.1 |
- |
4, 5 |
1135 |
4080970 |
BC068515.1 |
- |
4, 5 |
1171 |
4819377 |
View Full Product Details