Gene Transcripts

Gene Symbol
EXOC2
Entrez Gene ID
55770
Gene Name
exocyst complex component 2
Gene Aliases
SEC5, SEC5L1, Sec5p
Chromosome Location
Chr. 6: 485138 - 693141 on Build GRCh38
UniGene ID
Hs.484412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018303.5 NP_060773.3 28 2909
NR_073064.1 30 3071
XM_017011018.1 XP_016866507.1 29 2955
XM_017011019.1 XP_016866508.1 29 3117
XM_017011020.1 XP_016866509.1 29 2977
XM_017011021.1 XP_016866510.1 28 3213
XM_017011022.1 XP_016866511.1 28 2909
XM_017011023.1 XP_016866512.1 28 2909
XM_017011024.1 XP_016866513.1 29 2955
GenBank mRNA AJ414403.1 - 28 2833
AJ420556.1 - 6 428
AK001888.1 - 24 2324
AK056564.1 - 28 2877
AK302941.1 - 18 1650
AK314628.1 - 28 2909
AL833209.1 - 26 2465
AL833213.1 - 28 2855
BC016918.1 - 25 2389
BC080542.1 - 30 3041 5479496

View Full Product Details