Gene Transcripts

Gene Symbol
ST6GALNAC1
Entrez Gene ID
55808
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1
Gene Aliases
HSY11339, SIAT7A, ST6GalNAcI, STYI
Chromosome Location
Chr. 17: 76617769 - 76643838 on Build GRCh38
UniGene ID
Hs.105352
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289107.1 NP_001276036.1 7 1714
NM_018414.4 NP_060884.1 6 1577
XM_011524995.2 XP_011523297.1 6 1497
XM_011524996.2 XP_011523298.1 6 1497
XM_017024842.1 XP_016880331.1 6 1497
GenBank mRNA AK000113.1 - 5 731
AY096001.1 - 6 1377
AY358918.1 - 6 1522
Y11339.2 - 6 1437

View Full Product Details