Gene Transcripts
- Gene Symbol
- PSMA7
- Entrez Gene ID
- 5688
- Gene Name
- proteasome subunit alpha 7
- Gene Aliases
- C6, HEL-S-276, HSPC, RC6-1, XAPC7
- Chromosome Location
- Chr. 20: 62136727 - 62143458 on Build GRCh38
- UniGene ID
- Hs.233952
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002792.3 |
NP_002783.1
|
5, 6 |
753 |
|
GenBank mRNA |
AF022815.1 |
- |
5, 6 |
622 |
|
AF054185.1 |
- |
5, 6 |
683 |
|
AK312025.1 |
- |
5, 6 |
648 |
|
BC001895.1 |
- |
5, 6 |
686 |
|
BC004427.1 |
- |
5, 6 |
671 |
2819923 |
BC014999.1 |
- |
5, 6 |
681 |
|
BT007165.1 |
- |
5, 6 |
598 |
|
BU632089.1 |
- |
2, 3 |
292 |
|
BU687782.1 |
- |
2, 3 |
294 |
|
GQ472224.1 |
- |
5, 6 |
671 |
|
View Full Product Details