Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300942.1 | NP_001287871.1 | 16 | 2496 | |
NM_001300943.1 | NP_001287872.1 | 15 | 2454 | ||
NM_001300944.1 | NP_001287873.1 | 16 | 2496 | ||
NM_020193.4 | NP_064578.2 | 15 | 2451 | ||
XM_005274112.4 | XP_005274169.1 | 15 | 2440 | ||
XM_006718638.3 | XP_006718701.1 | 15 | 2464 | ||
XM_011545173.2 | XP_011543475.1 | 16 | 2593 | ||
XM_011545174.2 | XP_011543476.1 | 16 | 2590 | ||
XM_011545175.2 | XP_011543477.1 | 15 | 2464 | ||
XM_011545177.2 | XP_011543479.1 | 15 | 2440 | ||
XM_011545179.2 | XP_011543481.1 | 14 | 2311 | ||
XM_017018021.1 | XP_016873510.1 | 16 | 2593 | ||
XM_017018022.1 | XP_016873511.1 | 16 | 2439 | ||
GenBank mRNA | AK125114.1 | - | 14 | 2165 | |
AK126030.1 | - | 3 | 470 | ||
AK304106.1 | - | 3 | 361 | ||
BC117265.1 | - | 14 | 2350 | 40125816 | |
BC143374.1 | - | 15 | 2392 | 9051879 | |
BC143376.1 | - | 15 | 2392 | 9051881 | |
BX647398.1 | - | 4 | 546 |