Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004578.3 | NP_004569.2 | 5 | 581 | |
GenBank mRNA | AF498934.1 | - | 5 | 303 | |
AK223009.1 | - | 5 | 346 | ||
AK313807.1 | - | 5 | 554 | ||
AY585832.1 | - | 5 | 510 | ||
BC002438.1 | - | 5 | 374 | 3346455 | |
BC004309.1 | - | 5 | 511 | 3627110 | |
M28211.1 | - | 5 | 358 | ||
X13902.1 | - | 5 | 414 |