Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002898.3 | NP_002889.1 | 10 | 1120 | |
XM_005269059.4 | XP_005269116.1 | 10 | 988 | ||
XM_005269060.4 | XP_005269117.1 | 10 | 988 | ||
XM_005269061.3 | XP_005269118.1 | 10 | 988 | ||
XM_006719541.3 | XP_006719604.1 | 10 | 988 | ||
XM_006719542.3 | XP_006719605.1 | 10 | 988 | ||
XM_006719543.3 | XP_006719606.1 | 10 | 988 | ||
XM_006719544.3 | XP_006719607.1 | 10 | 988 | ||
XM_011538637.2 | XP_011536939.1 | 10 | 932 | ||
XM_011538638.2 | XP_011536940.1 | 11 | 1254 | ||
XM_011538639.2 | XP_011536941.1 | 10 | 988 | ||
XM_011538640.2 | XP_011536942.1 | 10 | 988 | ||
XM_011538642.2 | XP_011536944.1 | 8 | 718 | ||
XM_017019775.1 | XP_016875264.1 | 10 | 927 | ||
XM_017019776.1 | XP_016875265.1 | 6 | 412 | ||
XM_017019777.1 | XP_016875266.1 | 9 | 836 | ||
XM_017019778.1 | XP_016875267.1 | 6 | 428 | ||
GenBank mRNA | AK293315.1 | - | 9 | 720 | |
AK299580.1 | - | 7 | 579 | ||
AK304540.1 | - | 9 | 780 | ||
AK308885.1 | - | 8 | 816 | ||
BC027863.1 | - | 10 | 929 | 5202634 | |
BC072679.1 | - | 10 | 993 | 6048623 | |
D28483.1 | - | 10 | 937 |