Gene Transcripts

Gene Symbol
RP2
Entrez Gene ID
6102
Gene Name
retinitis pigmentosa 2 (X-linked recessive)
Gene Aliases
DELXp11.3, NM23-H10, NME10, TBCCD2, XRP2
Chromosome Location
Chr. X: 46836912 - 46882358 on Build GRCh38
UniGene ID
Hs.44766
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006915.2 NP_008846.2 2 506
GenBank mRNA AJ007590.1 - 2 489
AK312555.1 - 2 340
BC043348.2 - 2 428 5520795
BC053530.1 - 2 426 3871941
DA211161.1 - 2 507

View Full Product Details