Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021630.5 | NP_067643.3 | 3 | 932 | |
NM_176871.3 | NP_789847.1 | 2 | 274 | ||
NM_198042.3 | NP_932159.1 | 2 | 274 | ||
GenBank mRNA | AF289563.1 | - | 3 | 531 | |
AK024498.1 | - | 2 | 217 | ||
AK027800.1 | - | 3 | 543 | ||
AK092968.1 | - | 3 | 932 | ||
AY007729.1 | - | 2 | 298 | ||
AY070438.1 | - | 2 | 298 | ||
AY217349.1 | - | 2 | 259 | ||
BC021556.1 | - | 3 | 413 | 3959372 | |
BC071774.1 | - | 3 | 543 | 4593207 | |
DN997519.1 | - | 3 | 218 |