Gene Transcripts

Gene Symbol
PIRT
Entrez Gene ID
644139
Gene Name
phosphoinositide interacting regulator of transient receptor potential channels
Gene Aliases
Chromosome Location
Chr. 17: 10822475 - 10838101 on Build GRCh38
UniGene ID
Hs.553909
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101387.1 NP_001094857.1 2 325
GenBank mRNA AK021549.1 - 2 306
AK299808.1 - 2 311
DC332889.1 - 2 325

View Full Product Details