Gene Transcripts

Gene Symbol
STARD9
Entrez Gene ID
57519
Gene Name
StAR related lipid transfer domain containing 9
Gene Aliases
KIF16A
Chromosome Location
Chr. 15: 42575646 - 42720998 on Build GRCh38
UniGene ID
Hs.122061
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020759.2 NP_065810.2 7, 8 613
XM_011521831.2 XP_011520133.1 7, 8 626
XM_011521832.2 XP_011520134.1 7, 8 623
XM_011521833.2 XP_011520135.1 6, 7 593
XM_011521834.2 XP_011520136.1 7, 8 626
XM_011521835.2 XP_011520137.1 7, 8 626
XM_011521836.2 XP_011520138.1 7, 8 928
XM_011521837.2 XP_011520139.1 5, 6 395
XM_017022439.1 XP_016877928.1 7, 8 626
XM_017022440.1 XP_016877929.1 6, 7 525
XM_017022441.1 XP_016877930.1 5, 6 844
XM_017022442.1 XP_016877931.1 1, 2 20
GenBank mRNA AK122666.1 - 5, 6 627
CR627426.1 - 7, 8 613

View Full Product Details