Gene Transcripts

Gene Symbol
SBK2
Entrez Gene ID
646643
Gene Name
SH3 domain binding kinase family member 2
Gene Aliases
SGK069
Chromosome Location
Chr. 19: 55529336 - 55537175 on Build GRCh38
UniGene ID
Hs.532676
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101401.2 NP_001094871.2 1 249
XM_006723327.3 XP_006723390.1 2 358
XM_011527227.2 XP_011525529.1 2 357

View Full Product Details