Gene Transcripts

Gene Symbol
SKOR2
Entrez Gene ID
652991
Gene Name
SKI family transcriptional corepressor 2
Gene Aliases
CH18515, CORL2, FUSSEL18
Chromosome Location
Chr. 18: 47206182 - 47249524 on Build GRCh38
UniGene ID
Hs.585834
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278063.1 NP_001264992.1 1 2461
XM_006722525.3 XP_006722588.1 1 2802

View Full Product Details