Gene Transcripts
- Gene Symbol
- UFD1L
- Entrez Gene ID
- 7353
- Gene Name
- ubiquitin fusion degradation 1 like (yeast)
- Gene Aliases
- UFD1
- Chromosome Location
- Chr. 22: 19449941 - 19479215 on Build GRCh38
- UniGene ID
- Hs.474213
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001035247.2 |
NP_001030324.2
|
4, 5 |
424 |
|
NM_005659.6 |
NP_005650.2
|
4, 5 |
424 |
|
XM_011530361.2 |
XP_011528663.1
|
4, 5 |
550 |
|
GenBank mRNA |
AF141201.1 |
- |
3, 4 |
294 |
|
AJ239058.1 |
- |
3, 4 |
294 |
|
AK225877.1 |
- |
4, 5 |
381 |
|
AK304734.1 |
- |
4, 5 |
394 |
|
AK308786.1 |
- |
4, 5 |
368 |
|
AK310950.1 |
- |
4, 5 |
376 |
|
AK315998.1 |
- |
4, 5 |
375 |
|
AY101594.1 |
- |
4, 5 |
351 |
|
BC001049.2 |
- |
4, 5 |
371 |
3507963 |
BC005087.1 |
- |
4, 5 |
372 |
3830774 |
BM761958.1 |
- |
3, 4 |
363 |
|
BX648483.1 |
- |
4, 5 |
356 |
|
CR456607.1 |
- |
4, 5 |
308 |
|
DB496066.1 |
- |
4, 5 |
408 |
|
U64444.1 |
- |
4, 5 |
372 |
|
View Full Product Details