Gene Transcripts

Gene Symbol
PSAPL1
Entrez Gene ID
768239
Gene Name
prosaposin-like 1 (gene/pseudogene)
Gene Aliases
Chromosome Location
Chr. 4: 7430294 - 7434973 on Build GRCh38
UniGene ID
Hs.662523
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085382.1 NP_001078851.1 1 408
GenBank mRNA BC034552.1 - 1 323
BC068579.1 - 1 382
DQ991252.1 - 1 408

View Full Product Details