Gene Transcripts

Gene Symbol
RERGL
Entrez Gene ID
79785
Gene Name
RERG like
Gene Aliases
Chromosome Location
Chr. 12: 18080869 - 18090193 on Build GRCh38
UniGene ID
Hs.732831
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286201.1 NP_001273130.1 4 315
NM_024730.3 NP_079006.1 5 472
GenBank mRNA AK026308.1 - 5 398
BC042888.1 - 5 459 5198817
BX422875.2 - 4 263
DT220095.1 - 5 472

View Full Product Details