Gene Transcripts
- Gene Symbol
- MAP9
- Entrez Gene ID
- 79884
- Gene Name
- microtubule associated protein 9
- Gene Aliases
- ASAP
- Chromosome Location
- Chr. 4: 155342658 - 155377118 on Build GRCh38
- UniGene ID
- Hs.61271
- Species
- Homo sapiens
- Gene Symbol
- LOC102724776
- Entrez Gene ID
- 102724776
- Gene Name
- uncharacterized LOC102724776
- Gene Aliases
- Chromosome Location
- Chr. 4: 155354216 - 155360461 on Build GRCh38
- UniGene ID
- Hs.61271
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_125937.1 |
|
3 |
286 |
|
GenBank mRNA |
AA968415.1 |
- |
1 |
164 |
|
View Full Product Details