Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271895.1 | NP_001258824.1 | 12 | 1498 | |
NM_001271896.1 | NP_001258825.1 | 10 | 1354 | ||
NM_025150.4 | NP_079426.2 | 14 | 1744 | ||
NR_073513.1 | 10 | 1143 | |||
NR_073514.1 | 13 | 1619 | |||
XM_006711555.1 | XP_006711618.1 | 14 | 1707 | ||
XM_006711556.1 | XP_006711619.1 | 13 | 1560 | ||
XM_017002394.1 | XP_016857883.1 | 13 | 1560 | ||
XM_017002395.1 | XP_016857884.1 | 12 | 1461 | ||
GenBank mRNA | AK022590.1 | - | 10 | 1304 | |
AK222814.1 | - | 14 | 1685 | ||
AK225368.1 | - | 12 | 1418 | ||
AK315507.1 | - | 12 | 1444 | ||
AL512732.1 | - | 3 | 208 | ||
AY450558.1 | - | 10 | 1073 | ||
BC000541.2 | - | 14 | 1691 | 3163506 | |
BC007824.2 | - | 14 | 1632 | ||
BC009997.2 | - | 14 | 1664 | 4128881 |