Gene Transcripts

Gene Symbol
COIL
Entrez Gene ID
8161
Gene Name
coilin
Gene Aliases
CLN80, p80-coilin
Chromosome Location
Chr. 17: 56938199 - 56961050 on Build GRCh38
UniGene ID
Hs.532795
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004645.2 NP_004636.1 2 402
GenBank mRNA AK313616.1 - 2 384
BC010385.1 - 2 399 4275993
U06632.1 - 2 393

View Full Product Details