Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031923.3 | NP_114129.1 | 3 | 795 | |
XM_011519741.1 | XP_011518043.1 | 3 | 819 | ||
GenBank mRNA | AJ292190.1 | - | 3 | 589 | |
AK307521.1 | - | 3 | 768 | ||
AK307657.1 | - | 3 | 773 | ||
AL117661.1 | - | 3 | 523 | ||
BC017320.1 | - | 3 | 781 | ||
BC028077.1 | - | 3 | 752 | ||
BC045106.1 | - | 3 | 781 | ||
BC062352.1 | - | 3 | 762 | ||
BC073884.1 | - | 3 | 786 | ||
BC105593.1 | - | 3 | 799 |