Gene Transcripts
- Gene Symbol
- RAB11FIP4
- Entrez Gene ID
- 84440
- Gene Name
- RAB11 family interacting protein 4
- Gene Aliases
- RAB11-FIP4
- Chromosome Location
- Chr. 17: 31391624 - 31538217 on Build GRCh38
- UniGene ID
- Hs.406788
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001303542.2 |
NP_001290471.2
|
8, 9 |
1218 |
|
NM_032932.5 |
NP_116321.2
|
10, 11 |
1511 |
|
GenBank mRNA |
AB058724.1 |
- |
10, 11 |
1200 |
|
AJ314646.2 |
- |
10, 11 |
1200 |
|
AK074341.1 |
- |
3, 4 |
289 |
|
AK097424.1 |
- |
8, 9 |
1161 |
|
AL831830.1 |
- |
2, 3 |
260 |
|
AY169244.1 |
- |
10, 11 |
1511 |
|
BC093914.1 |
- |
10, 11 |
1314 |
7939759 |
BC101517.1 |
- |
10, 11 |
1314 |
8069023 |
View Full Product Details