Gene Transcripts

Gene Symbol
GPT2
Entrez Gene ID
84706
Gene Name
glutamic--pyruvic transaminase 2
Gene Aliases
ALT2, GPT 2, MRT49
Chromosome Location
Chr. 16: 46884380 - 46931297 on Build GRCh38
UniGene ID
Hs.460693
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133443.3 NP_597700.1 2 269
GenBank mRNA AY029173.2 - 2 253
BC062555.1 - 2 218 6429257
HY117879.1 - 2 269

View Full Product Details