Gene Transcripts
- Gene Symbol
- CST7
- Entrez Gene ID
- 8530
- Gene Name
- cystatin F
- Gene Aliases
- CMAP
- Chromosome Location
- Chr. 20: 24949230 - 24959928 on Build GRCh38
- UniGene ID
- Hs.143212
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003650.3 |
NP_003641.3
|
2, 3 |
513 |
|
GenBank mRNA |
AB015225.1 |
- |
2, 3 |
385 |
|
AF031824.1 |
- |
2, 3 |
452 |
|
AF036342.1 |
- |
2, 3 |
294 |
|
BC015507.1 |
- |
2, 3 |
408 |
3893020 |
BG547285.1 |
- |
2, 3 |
514 |
|
BT009825.1 |
- |
2, 3 |
303 |
|
CA449286.1 |
- |
2, 3 |
424 |
|
CR541860.1 |
- |
2, 3 |
237 |
|
CR541878.1 |
- |
2, 3 |
237 |
|
View Full Product Details