Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003679.4 | NP_003670.2 | 13 | 1492 | |
XM_005273337.1 | XP_005273394.1 | 13 | 1563 | ||
XM_005273338.3 | XP_005273395.1 | 14 | 1845 | ||
XM_017002649.1 | XP_016858138.1 | 14 | 1836 | ||
XM_017002650.1 | XP_016858139.1 | 11 | 1260 | ||
GenBank mRNA | AF056032.1 | - | 13 | 1227 | |
AK291558.1 | - | 13 | 1248 | ||
AK310937.1 | - | 13 | 1369 | ||
BC005297.1 | - | 13 | 1237 | 3934714 | |
Y13153.1 | - | 13 | 1233 |