Gene Transcripts

Gene Symbol
GGACT
Entrez Gene ID
87769
Gene Name
gamma-glutamylamine cyclotransferase
Gene Aliases
A2LD1
Chromosome Location
Chr. 13: 100530164 - 100588792 on Build GRCh38
UniGene ID
Hs.350868
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195087.1 NP_001182016.1 3 579
NM_033110.2 NP_149101.1 2 594
XM_005254083.2 XP_005254140.1 2 721
XM_011521129.2 XP_011519431.1 3 3557
XM_017020804.1 XP_016876293.1 3 695
GenBank mRNA AK091974.1 - 3 267
AK095850.1 - 3 579
BC001077.2 - 2 535
BC004360.1 - 2 535

View Full Product Details