Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133369.2 | NP_588610.2 | 13 | 2351 | |
XM_006714927.1 | XP_006714990.1 | 13 | 2331 | ||
XM_006714928.1 | XP_006714991.1 | 14 | 2499 | ||
XM_011534686.1 | XP_011532988.1 | 14 | 2499 | ||
GenBank mRNA | AB075856.1 | - | 1 | 4213 | |
BC009333.2 | - | 7 | 1257 | ||
BC122556.1 | - | 7 | 1228 | ||
BC157824.1 | - | 13 | 2234 | 9057043 |