Gene Transcripts
- Gene Symbol
- MTG1
- Entrez Gene ID
- 92170
- Gene Name
- mitochondrial ribosome associated GTPase 1
- Gene Aliases
- GTP, GTPBP7
- Chromosome Location
- Chr. 10: 133394117 - 133420670 on Build GRCh38
- UniGene ID
- Hs.501578
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138384.2 |
NP_612393.2
|
10, 11 |
971 |
|
View Full Product Details