Gene Transcripts
- Gene Symbol
- BBIP1
- Entrez Gene ID
- 92482
- Gene Name
- BBSome interacting protein 1
- Gene Aliases
- BBIP10, BBS18, NCRNA00081, bA348N5.3
- Chromosome Location
- Chr. 10: 110898730 - 110919366 on Build GRCh38
- UniGene ID
- Hs.232543
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001195304.1 |
NP_001182233.1
|
4, 5 |
581 |
|
NM_001195305.1 |
NP_001182234.1
|
3, 4 |
424 |
|
NM_001195306.1 |
NP_001182235.1
|
3, 4 |
270 |
|
NM_001195307.1 |
NP_001182236.1
|
2, 3 |
349 |
|
NM_001243783.1 |
NP_001230712.1
|
2, 3 |
331 |
|
GenBank mRNA |
AK025724.1 |
- |
3, 4 |
244 |
|
AK125829.1 |
- |
1 |
4428 |
|
AK307126.1 |
- |
3, 4 |
270 |
|
AK307314.1 |
- |
4, 5 |
457 |
|
BC104248.1 |
- |
3, 4 |
231 |
|
BC104249.1 |
- |
3, 4 |
233 |
|
BC105804.1 |
- |
4, 5 |
420 |
|
BC113000.1 |
- |
3, 4 |
233 |
|
BC113001.1 |
- |
3, 4 |
233 |
|
BI552279.1 |
- |
2, 3 |
163 |
|
DB450918.1 |
- |
3, 4 |
424 |
|
View Full Product Details