Gene Transcripts

Gene Symbol
FBXO44
Entrez Gene ID
93611
Gene Name
F-box protein 44
Gene Aliases
FBG3, FBX30, FBX6A, Fbx44, Fbxo6a
Chromosome Location
Chr. 1: 11654375 - 11663327 on Build GRCh38
UniGene ID
Hs.556006
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014765.1 NP_001014765.1 4 790
NM_001304790.1 NP_001291719.1 4 448
NM_001304791.1 NP_001291720.1 3 529
NM_033182.5 NP_149438.2 3 418
NM_183412.2 NP_904319.1 3 529
NM_183413.2 NP_904320.1 3 418
NR_130909.1 3 284
XM_005263535.1 XP_005263592.1 3 389
XM_005263536.4 XP_005263593.1 4 783
XM_005263537.1 XP_005263594.1 3 529
XM_006711043.2 XP_006711106.1 3 389
XM_006711045.2 XP_006711108.1 3 391
XM_011542435.1 XP_011540737.1 4 851
XM_017002842.1 XP_016858331.1 3 389
XM_017002843.1 XP_016858332.1 3 391
XM_017002844.1 XP_016858333.1 3 389
GenBank mRNA AK055344.1 - 3 529
AK293721.1 - 3 366
AK294053.1 - 3 352
AK296025.1 - 3 264
AK301418.1 - 3 398
AK316284.1 - 3 398
AY007380.1 - 2 325
AY040878.1 - 3 374
BC007832.2 - 3 365 4054414
BM546141.1 - 3 379
BM903606.1 - 425
BU164151.1 - 4 372
HY152948.1 - 3 417

View Full Product Details