Gene Transcripts

Gene Symbol
RAB28
Entrez Gene ID
9364
Gene Name
RAB28, member RAS oncogene family
Gene Aliases
CORD18
Chromosome Location
Chr. 4: 13293479 - 13484365 on Build GRCh38
UniGene ID
Hs.656060
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017979.2 NP_001017979.1 2, 3 395
NM_001159601.1 NP_001153073.1 2, 3 395
NM_004249.3 NP_004240.2 2, 3 395
XM_005248218.4 XP_005248275.1 2, 3 395
XM_011513911.2 XP_011512213.1 2, 3 395
XM_011513912.2 XP_011512214.1 2, 3 1396
XM_017008850.1 XP_016864339.1 2, 3 395
GenBank mRNA AF498955.1 - 2, 3 180
BC018067.1 - 2, 3 361
BC035054.2 - 2, 3 368 5263482
CX165950.1 - 2, 3 215
DA369194.1 - 2, 3 395
X94703.1 - 2, 3 180

View Full Product Details