Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 95836055-95836461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR2
Entrez Gene ID
8898
Gene Name
myotubularin related protein 2
Gene Aliases
CMT4B, CMT4B1
UniGene
Hs.181326
Gene Chromosome Location
Chr.11: 95832880-95924207 on Build GRCh38
Transcripts
NM_001243571.1 NM_016156.5 NM_201278.2 NM_201281.2 XM_005274374.2 XM_005274375.2 XM_006718934.2 XM_006718935.2 XM_006718936.3 XM_011543058.2 XM_011543059.2 XM_017018517.1 XM_017018518.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1662874 c.1726T>C p.W576R Chr.11: 95836192-95836192 on Build GRCh38
1662875 c.1510T>C p.W504R Chr.11: 95836192-95836192 on Build GRCh38
2061853 c.1770+1G>A p.? Chr.11: 95836147-95836147 on Build GRCh38
2061854 c.1554+1G>A p.? Chr.11: 95836147-95836147 on Build GRCh38
6070742 c.1471G>A p.V491I Chr.11: 95836231-95836231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563383988 Chr.11: 95836422-95836422 on Build GRCh38
rs755619692 Chr.11: 95836130-95836130 on Build GRCh38
rs756909627 Chr.11: 95836145-95836145 on Build GRCh38
rs931675214 Chr.11: 95836277-95836277 on Build GRCh38
rs61735576 Chr.11: 95836161-95836161 on Build GRCh38
rs781368440 Chr.11: 95836288-95836288 on Build GRCh38
rs142687077 Chr.11: 95836300-95836300 on Build GRCh38
rs749533331 Chr.11: 95836321-95836321 on Build GRCh38
rs766722493 Chr.11: 95836239-95836239 on Build GRCh38
rs774911582 Chr.11: 95836242-95836242 on Build GRCh38
rs750032278 Chr.11: 95836356-95836356 on Build GRCh38
rs981384605 Chr.11: 95836411-95836411 on Build GRCh38
rs774704606 Chr.11: 95836333-95836333 on Build GRCh38
rs766890540 Chr.11: 95836126-95836126 on Build GRCh38
rs149476960 Chr.11: 95836177-95836177 on Build GRCh38
rs935486210 Chr.11: 95836290-95836290 on Build GRCh38
rs746873563 Chr.11: 95836283-95836283 on Build GRCh38
rs762227896 Chr.11: 95836294-95836294 on Build GRCh38
rs958180213 Chr.11: 95836128-95836139 on Build GRCh38
rs773652596 Chr.11: 95836105-95836105 on Build GRCh38
rs183056793 Chr.11: 95836374-95836374 on Build GRCh38
rs752540236 Chr.11: 95836252-95836253 on Build GRCh38
rs753377983 Chr.11: 95836249-95836249 on Build GRCh38
rs754673845 Chr.11: 95836423-95836423 on Build GRCh38
rs763654400 Chr.11: 95836132-95836132 on Build GRCh38
rs770074296 Chr.11: 95836209-95836209 on Build GRCh38
rs71473257 Chr.11: 95836334-95836334 on Build GRCh38
rs763315235 Chr.11: 95836114-95836114 on Build GRCh38
rs765627503 Chr.11: 95836101-95836101 on Build GRCh38
rs765914011 Chr.11: 95836105-95836106 on Build GRCh38
rs138626568 Chr.11: 95836217-95836217 on Build GRCh38
rs761381781 Chr.11: 95836346-95836346 on Build GRCh38
rs778884253 Chr.11: 95836327-95836328 on Build GRCh38
rs558018 Chr.11: 95836284-95836284 on Build GRCh38
rs777913101 Chr.11: 95836325-95836326 on Build GRCh38
rs573754936 Chr.11: 95836129-95836138 on Build GRCh38
rs147170889 Chr.11: 95836251-95836251 on Build GRCh38
rs754608173 Chr.11: 95836136-95836141 on Build GRCh38
rs894019754 Chr.11: 95836313-95836313 on Build GRCh38
rs771548803 Chr.11: 95836197-95836197 on Build GRCh38
rs574241508 Chr.11: 95836314-95836314 on Build GRCh38
rs963277227 Chr.11: 95836430-95836430 on Build GRCh38
rs627568 Chr.11: 95836089-95836089 on Build GRCh38
rs138036650 Chr.11: 95836163-95836163 on Build GRCh38
rs760091979 Chr.11: 95836353-95836353 on Build GRCh38
rs750110066 Chr.11: 95836257-95836257 on Build GRCh38
rs756113331 Chr.11: 95836323-95836324 on Build GRCh38
rs1053053925 Chr.11: 95836309-95836309 on Build GRCh38
rs763529973 Chr.11: 95836248-95836248 on Build GRCh38
rs1004694376 Chr.11: 95836409-95836409 on Build GRCh38
rs369303651 Chr.11: 95836133-95836133 on Build GRCh38
rs563549676 Chr.11: 95836140-95836141 on Build GRCh38
rs772590622 Chr.11: 95836343-95836343 on Build GRCh38
rs772191796 Chr.11: 95836328-95836329 on Build GRCh38
rs762336648 Chr.11: 95836374-95836375 on Build GRCh38
rs757375439 Chr.11: 95836326-95836327 on Build GRCh38
rs765931267 Chr.11: 95836095-95836096 on Build GRCh38
rs908222470 Chr.11: 95836141-95836141 on Build GRCh38
rs186569484 Chr.11: 95836082-95836082 on Build GRCh38
rs752050148 Chr.11: 95836128-95836128 on Build GRCh38
rs751182047 Chr.11: 95836129-95836134 on Build GRCh38
rs778532655 Chr.11: 95836148-95836148 on Build GRCh38
rs748212593 Chr.11: 95836298-95836298 on Build GRCh38
rs780983110 Chr.11: 95836141-95836142 on Build GRCh38
rs376530848 Chr.11: 95836281-95836281 on Build GRCh38
rs545964321 Chr.11: 95836398-95836398 on Build GRCh38
rs777164308 Chr.11: 95836330-95836331 on Build GRCh38
rs201996397 Chr.11: 95836344-95836344 on Build GRCh38
rs114259367 Chr.11: 95836369-95836369 on Build GRCh38
rs61735577 Chr.11: 95836162-95836162 on Build GRCh38
rs760050731 Chr.11: 95836245-95836245 on Build GRCh38
rs771272314 Chr.11: 95836332-95836332 on Build GRCh38
rs773359524 Chr.11: 95836211-95836211 on Build GRCh38
rs574344138 Chr.11: 95836130-95836141 on Build GRCh38
rs748336978 Chr.11: 95836207-95836207 on Build GRCh38
rs9919542 Chr.11: 95836366-95836366 on Build GRCh38
rs200717440 Chr.11: 95836224-95836224 on Build GRCh38
rs764911369 Chr.11: 95836252-95836252 on Build GRCh38

View Full Product Details