Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102942941-102943480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP13
Entrez Gene ID
4322
Gene Name
matrix metallopeptidase 13
Gene Aliases
CLG3, MANDP1, MMP-13
UniGene
Hs.2936
Gene Chromosome Location
Chr.11: 102942992-102955734 on Build GRCh38
Transcripts
NM_002427.3

SNP Detail

SNP ID Chromosome Location
rs886047551 Chr.11: 102943259-102943259 on Build GRCh38
rs533082843 Chr.11: 102943232-102943232 on Build GRCh38
rs782349637 Chr.11: 102943037-102943037 on Build GRCh38
rs782048471 Chr.11: 102943371-102943371 on Build GRCh38
rs137968508 Chr.11: 102943218-102943218 on Build GRCh38
rs530128848 Chr.11: 102943189-102943189 on Build GRCh38
rs781904671 Chr.11: 102943251-102943252 on Build GRCh38
rs17860586 Chr.11: 102943202-102943202 on Build GRCh38
rs184593720 Chr.11: 102943045-102943045 on Build GRCh38
rs938587284 Chr.11: 102943411-102943411 on Build GRCh38
rs782016341 Chr.11: 102943387-102943387 on Build GRCh38
rs1026730658 Chr.11: 102943196-102943196 on Build GRCh38
rs184451705 Chr.11: 102943129-102943129 on Build GRCh38
rs1042840 Chr.11: 102943381-102943381 on Build GRCh38
rs113572120 Chr.11: 102943128-102943128 on Build GRCh38
rs542211271 Chr.11: 102942991-102942991 on Build GRCh38
rs903845493 Chr.11: 102943108-102943108 on Build GRCh38
rs1018380265 Chr.11: 102943164-102943164 on Build GRCh38
rs886047552 Chr.11: 102943416-102943417 on Build GRCh38
rs1008915193 Chr.11: 102943142-102943142 on Build GRCh38
rs188511209 Chr.11: 102943047-102943047 on Build GRCh38
rs181285292 Chr.11: 102943066-102943066 on Build GRCh38
rs17860587 Chr.11: 102943125-102943125 on Build GRCh38
rs886047550 Chr.11: 102943164-102943164 on Build GRCh38
rs532528718 Chr.11: 102943414-102943414 on Build GRCh38
rs35477433 Chr.11: 102943268-102943269 on Build GRCh38
rs964209491 Chr.11: 102943175-102943175 on Build GRCh38
rs915733663 Chr.11: 102943450-102943450 on Build GRCh38
rs576331724 Chr.11: 102942974-102942974 on Build GRCh38

View Full Product Details