Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112086758-112087260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDHD
Entrez Gene ID
6392
Gene Name
succinate dehydrogenase complex subunit D
Gene Aliases
CBT1, CII-4, CWS3, PGL, PGL1, QPs3, SDH4, cybS
UniGene
Hs.356270
Gene Chromosome Location
Chr.11: 112086824-112095801 on Build GRCh38
Transcripts
NM_001276503.1 NM_001276504.1 NM_001276506.1 NM_003002.3 NR_077060.1 NM_012459.2 NR_028383.1

Gene Symbol
TIMM8B
Entrez Gene ID
26521
Gene Name
translocase of inner mitochondrial membrane 8 homolog B
Gene Aliases
DDP2, TIM8B
UniGene
Hs.279915
Gene Chromosome Location
Chr.11: 112084800-112086798 on Build GRCh38
Transcripts
NM_001276503.1 NM_001276504.1 NM_001276506.1 NM_003002.3 NR_077060.1 NM_012459.2 NR_028383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1351278 c.10C>T p.L4F Chr.11: 112086917-112086917 on Build GRCh38
6953300 c.34G>A p.G12S Chr.11: 112086941-112086941 on Build GRCh38
6912550 c.41T>G p.L14R Chr.11: 112086948-112086948 on Build GRCh38
7003838 c.2T>C p.M1T Chr.11: 112086767-112086767 on Build GRCh38
4967348 c.15G>T p.W5C Chr.11: 112086922-112086922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs587776644 Chr.11: 112086961-112086961 on Build GRCh38
rs780972546 Chr.11: 112086899-112086899 on Build GRCh38
rs564402774 Chr.11: 112086827-112086827 on Build GRCh38
rs999780639 Chr.11: 112087170-112087170 on Build GRCh38
rs752724822 Chr.11: 112086835-112086835 on Build GRCh38
rs544421571 Chr.11: 112086826-112086826 on Build GRCh38
rs767806458 Chr.11: 112086878-112086878 on Build GRCh38
rs878854593 Chr.11: 112086949-112086949 on Build GRCh38
rs940936212 Chr.11: 112086944-112086944 on Build GRCh38
rs573701234 Chr.11: 112087141-112087141 on Build GRCh38
rs763548563 Chr.11: 112087030-112087030 on Build GRCh38
rs954757882 Chr.11: 112087236-112087236 on Build GRCh38
rs748406508 Chr.11: 112086808-112086808 on Build GRCh38
rs12789541 Chr.11: 112087182-112087182 on Build GRCh38
rs1003347831 Chr.11: 112086776-112086776 on Build GRCh38
rs568087116 Chr.11: 112087129-112087129 on Build GRCh38
rs996198487 Chr.11: 112087213-112087213 on Build GRCh38
rs1029054137 Chr.11: 112087224-112087224 on Build GRCh38
rs995872549 Chr.11: 112087199-112087199 on Build GRCh38
rs104894307 Chr.11: 112086908-112086908 on Build GRCh38
rs11550094 Chr.11: 112086931-112086931 on Build GRCh38
rs763197227 Chr.11: 112086857-112086857 on Build GRCh38
rs200704155 Chr.11: 112086883-112086883 on Build GRCh38
rs200665443 Chr.11: 112086853-112086853 on Build GRCh38
rs80338842 Chr.11: 112086910-112086910 on Build GRCh38
rs954474648 Chr.11: 112086972-112086972 on Build GRCh38
rs766792211 Chr.11: 112087109-112087109 on Build GRCh38
rs968741221 Chr.11: 112086832-112086832 on Build GRCh38
rs201097536 Chr.11: 112086861-112086861 on Build GRCh38
rs989438518 Chr.11: 112086799-112086799 on Build GRCh38
rs764936238 Chr.11: 112086887-112086887 on Build GRCh38
rs1008619752 Chr.11: 112087219-112087219 on Build GRCh38
rs764384503 Chr.11: 112086942-112086942 on Build GRCh38
rs542296302 Chr.11: 112086775-112086775 on Build GRCh38
rs555586706 Chr.11: 112086784-112086784 on Build GRCh38
rs778202647 Chr.11: 112086920-112086920 on Build GRCh38
rs753152376 Chr.11: 112086881-112086881 on Build GRCh38
rs866893113 Chr.11: 112086793-112086793 on Build GRCh38
rs558914063 Chr.11: 112086930-112086930 on Build GRCh38
rs974401612 Chr.11: 112086928-112086928 on Build GRCh38
rs894056577 Chr.11: 112087190-112087190 on Build GRCh38
rs770924084 Chr.11: 112086924-112086924 on Build GRCh38
rs200386359 Chr.11: 112086998-112086998 on Build GRCh38
rs902647432 Chr.11: 112087164-112087164 on Build GRCh38
rs757898976 Chr.11: 112086778-112086778 on Build GRCh38
rs973261024 Chr.11: 112087065-112087065 on Build GRCh38
rs772671893 Chr.11: 112086932-112086932 on Build GRCh38
rs375890857 Chr.11: 112086900-112086900 on Build GRCh38
rs931347289 Chr.11: 112087084-112087084 on Build GRCh38
rs1047012321 Chr.11: 112087158-112087158 on Build GRCh38
rs11547889 Chr.11: 112086923-112086923 on Build GRCh38
rs201382577 Chr.11: 112086889-112086889 on Build GRCh38
rs774734438 Chr.11: 112086868-112086868 on Build GRCh38
rs547875751 Chr.11: 112087194-112087194 on Build GRCh38
rs1014198262 Chr.11: 112086812-112086812 on Build GRCh38
rs999747150 Chr.11: 112086892-112086892 on Build GRCh38
rs878854589 Chr.11: 112086917-112086918 on Build GRCh38
rs1054078454 Chr.11: 112087173-112087173 on Build GRCh38
rs760995161 Chr.11: 112086885-112086885 on Build GRCh38
rs559368739 Chr.11: 112087070-112087070 on Build GRCh38
rs750080041 Chr.11: 112086945-112086945 on Build GRCh38
rs1022043662 Chr.11: 112086823-112086823 on Build GRCh38
rs867238579 Chr.11: 112087149-112087149 on Build GRCh38
rs540552714 Chr.11: 112086904-112086904 on Build GRCh38
rs200063990 Chr.11: 112086873-112086873 on Build GRCh38
rs200895313 Chr.11: 112086925-112086925 on Build GRCh38
rs916675733 Chr.11: 112087120-112087120 on Build GRCh38
rs200968062 Chr.11: 112086983-112086983 on Build GRCh38
rs770193469 Chr.11: 112086811-112086811 on Build GRCh38
rs968668917 Chr.11: 112087209-112087209 on Build GRCh38
rs34677591 Chr.11: 112086941-112086941 on Build GRCh38
rs956823778 Chr.11: 112086796-112086796 on Build GRCh38
rs765826714 Chr.11: 112086970-112086970 on Build GRCh38
rs1003320468 Chr.11: 112087121-112087121 on Build GRCh38
rs199955367 Chr.11: 112086896-112086896 on Build GRCh38
rs202196763 Chr.11: 112086991-112086991 on Build GRCh38
rs199890548 Chr.11: 112086958-112086958 on Build GRCh38
rs202003728 Chr.11: 112086858-112086858 on Build GRCh38
rs148364534 Chr.11: 112086789-112086789 on Build GRCh38
rs757913227 Chr.11: 112086893-112086893 on Build GRCh38
rs201556521 Chr.11: 112087005-112087005 on Build GRCh38
rs370761122 Chr.11: 112086967-112086967 on Build GRCh38
rs961948952 Chr.11: 112087054-112087056 on Build GRCh38
rs104894310 Chr.11: 112086921-112086921 on Build GRCh38
rs104894309 Chr.11: 112086940-112086940 on Build GRCh38
rs1032016970 Chr.11: 112086917-112086917 on Build GRCh38
rs933452204 Chr.11: 112086897-112086897 on Build GRCh38
rs773541015 Chr.11: 112086818-112086818 on Build GRCh38
rs751103729 Chr.11: 112087049-112087049 on Build GRCh38
rs7121782 Chr.11: 112087095-112087095 on Build GRCh38
rs372894110 Chr.11: 112086888-112086888 on Build GRCh38
rs780997752 Chr.11: 112086807-112086807 on Build GRCh38
rs754260613 Chr.11: 112087191-112087191 on Build GRCh38

View Full Product Details