Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52446954-52447464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT6B
Entrez Gene ID
3854
Gene Name
keratin 6B
Gene Aliases
CK-6B, CK6B, K6B, KRTL1, PC2, PC4
UniGene
Hs.708950
Gene Chromosome Location
Chr.12: 52446651-52452126 on Build GRCh38
Transcripts
NM_005555.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3462264 c.1527G>A p.L509L Chr.12: 52447358-52447358 on Build GRCh38
3462265 c.1518C>T p.G506G Chr.12: 52447367-52447367 on Build GRCh38
3462262 c.1677G>A p.R559R Chr.12: 52447208-52447208 on Build GRCh38
6698089 c.1573G>T p.G525C Chr.12: 52447312-52447312 on Build GRCh38
3462263 c.1662C>T p.T554T Chr.12: 52447223-52447223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748185532 Chr.12: 52447193-52447193 on Build GRCh38
rs750303987 Chr.12: 52447304-52447304 on Build GRCh38
rs141409814 Chr.12: 52447245-52447245 on Build GRCh38
rs535888045 Chr.12: 52447003-52447003 on Build GRCh38
rs775142460 Chr.12: 52447230-52447230 on Build GRCh38
rs780534493 Chr.12: 52447348-52447348 on Build GRCh38
rs138933209 Chr.12: 52447220-52447220 on Build GRCh38
rs141150690 Chr.12: 52447314-52447314 on Build GRCh38
rs769329834 Chr.12: 52447281-52447282 on Build GRCh38
rs762618910 Chr.12: 52447232-52447232 on Build GRCh38
rs139358472 Chr.12: 52447362-52447362 on Build GRCh38
rs757924059 Chr.12: 52447173-52447173 on Build GRCh38
rs376429278 Chr.12: 52447309-52447309 on Build GRCh38
rs371758517 Chr.12: 52447206-52447206 on Build GRCh38
rs1052448705 Chr.12: 52447122-52447122 on Build GRCh38
rs770190928 Chr.12: 52447279-52447279 on Build GRCh38
rs143946299 Chr.12: 52447291-52447291 on Build GRCh38
rs758927616 Chr.12: 52447177-52447178 on Build GRCh38
rs763683330 Chr.12: 52447238-52447238 on Build GRCh38
rs771328301 Chr.12: 52447212-52447212 on Build GRCh38
rs755876853 Chr.12: 52447250-52447250 on Build GRCh38
rs145254155 Chr.12: 52447376-52447376 on Build GRCh38
rs920531074 Chr.12: 52447271-52447271 on Build GRCh38
rs546061164 Chr.12: 52447373-52447373 on Build GRCh38
rs568087038 Chr.12: 52447074-52447074 on Build GRCh38
rs148044538 Chr.12: 52447418-52447418 on Build GRCh38
rs61746354 Chr.12: 52447395-52447395 on Build GRCh38
rs777542750 Chr.12: 52447283-52447285 on Build GRCh38
rs777803130 Chr.12: 52447270-52447270 on Build GRCh38
rs749680153 Chr.12: 52447415-52447415 on Build GRCh38
rs755323118 Chr.12: 52447416-52447416 on Build GRCh38
rs867923192 Chr.12: 52447412-52447412 on Build GRCh38
rs764131660 Chr.12: 52447174-52447174 on Build GRCh38
rs763718347 Chr.12: 52447431-52447431 on Build GRCh38
rs146988096 Chr.12: 52447252-52447252 on Build GRCh38
rs749563252 Chr.12: 52447181-52447181 on Build GRCh38
rs767365375 Chr.12: 52447299-52447299 on Build GRCh38
rs748722642 Chr.12: 52447433-52447433 on Build GRCh38
rs200055232 Chr.12: 52447336-52447336 on Build GRCh38
rs200690379 Chr.12: 52447180-52447180 on Build GRCh38
rs373670186 Chr.12: 52447439-52447439 on Build GRCh38
rs746223280 Chr.12: 52447278-52447278 on Build GRCh38
rs575779389 Chr.12: 52447253-52447253 on Build GRCh38
rs147008947 Chr.12: 52447408-52447408 on Build GRCh38
rs554596962 Chr.12: 52447281-52447281 on Build GRCh38
rs747495865 Chr.12: 52447208-52447208 on Build GRCh38
rs145923896 Chr.12: 52447248-52447248 on Build GRCh38
rs752172491 Chr.12: 52447164-52447164 on Build GRCh38
rs138021918 Chr.12: 52447260-52447260 on Build GRCh38
rs746457511 Chr.12: 52447433-52447433 on Build GRCh38
rs770104486 Chr.12: 52447347-52447347 on Build GRCh38
rs749628792 Chr.12: 52447359-52447359 on Build GRCh38
rs973202585 Chr.12: 52447019-52447019 on Build GRCh38
rs61749468 Chr.12: 52447337-52447337 on Build GRCh38
rs548270201 Chr.12: 52447067-52447067 on Build GRCh38
rs760989336 Chr.12: 52447121-52447121 on Build GRCh38
rs201073484 Chr.12: 52447409-52447409 on Build GRCh38
rs544756853 Chr.12: 52446978-52446978 on Build GRCh38
rs892707747 Chr.12: 52447135-52447135 on Build GRCh38
rs750932643 Chr.12: 52447401-52447401 on Build GRCh38
rs771525357 Chr.12: 52447427-52447427 on Build GRCh38
rs150021015 Chr.12: 52447375-52447375 on Build GRCh38
rs61746355 Chr.12: 52447390-52447390 on Build GRCh38
rs777924533 Chr.12: 52447320-52447320 on Build GRCh38
rs774086396 Chr.12: 52447363-52447363 on Build GRCh38
rs148209422 Chr.12: 52447292-52447292 on Build GRCh38
rs146173634 Chr.12: 52447318-52447318 on Build GRCh38
rs750890523 Chr.12: 52447154-52447168 on Build GRCh38
rs140188440 Chr.12: 52447319-52447319 on Build GRCh38
rs567093999 Chr.12: 52447182-52447182 on Build GRCh38
rs958861782 Chr.12: 52447258-52447258 on Build GRCh38
rs747408258 Chr.12: 52447280-52447281 on Build GRCh38
rs766177343 Chr.12: 52447310-52447310 on Build GRCh38
rs757339399 Chr.12: 52447394-52447394 on Build GRCh38
rs200620417 Chr.12: 52447218-52447218 on Build GRCh38
rs747053952 Chr.12: 52447275-52447275 on Build GRCh38
rs539318306 Chr.12: 52447224-52447224 on Build GRCh38
rs146373607 Chr.12: 52447343-52447343 on Build GRCh38
rs751538324 Chr.12: 52447176-52447176 on Build GRCh38
rs954490591 Chr.12: 52447230-52447231 on Build GRCh38
rs900023373 Chr.12: 52447196-52447196 on Build GRCh38
rs759394434 Chr.12: 52447441-52447441 on Build GRCh38
rs781682935 Chr.12: 52447277-52447277 on Build GRCh38
rs542447561 Chr.12: 52447321-52447321 on Build GRCh38
rs759082678 Chr.12: 52447153-52447153 on Build GRCh38
rs147867894 Chr.12: 52447391-52447391 on Build GRCh38
rs772139797 Chr.12: 52447195-52447195 on Build GRCh38
rs776365127 Chr.12: 52447151-52447151 on Build GRCh38
rs762600737 Chr.12: 52447421-52447421 on Build GRCh38
rs948122084 Chr.12: 52447042-52447042 on Build GRCh38
rs940086820 Chr.12: 52447063-52447063 on Build GRCh38
rs373264955 Chr.12: 52447334-52447334 on Build GRCh38
rs149152547 Chr.12: 52447381-52447381 on Build GRCh38
rs753657782 Chr.12: 52447382-52447382 on Build GRCh38
rs762671974 Chr.12: 52447288-52447288 on Build GRCh38
rs757227143 Chr.12: 52447241-52447241 on Build GRCh38
rs149495966 Chr.12: 52447276-52447276 on Build GRCh38
rs1048350395 Chr.12: 52447026-52447026 on Build GRCh38
rs75893729 Chr.12: 52447080-52447080 on Build GRCh38
rs770916621 Chr.12: 52447435-52447435 on Build GRCh38
rs909900755 Chr.12: 52447032-52447032 on Build GRCh38
rs780343812 Chr.12: 52447274-52447275 on Build GRCh38
rs751594384 Chr.12: 52447240-52447240 on Build GRCh38
rs779143449 Chr.12: 52447190-52447190 on Build GRCh38
rs549184060 Chr.12: 52447162-52447162 on Build GRCh38

View Full Product Details