Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52449451-52449944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT6B
Entrez Gene ID
3854
Gene Name
keratin 6B
Gene Aliases
CK-6B, CK6B, K6B, KRTL1, PC2, PC4
UniGene
Hs.708950
Gene Chromosome Location
Chr.12: 52446651-52452126 on Build GRCh38
Transcripts
NM_005555.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6698090 c.976C>T p.R326C Chr.12: 52449570-52449570 on Build GRCh38
1362494 c.871C>A p.L291I Chr.12: 52449799-52449799 on Build GRCh38
5984155 c.927G>A p.M309I Chr.12: 52449619-52449619 on Build GRCh38
394627 c.873T>C p.L291L Chr.12: 52449797-52449797 on Build GRCh38
3952889 c.1059G>A p.E353E Chr.12: 52449487-52449487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781042956 Chr.12: 52449505-52449505 on Build GRCh38
rs756807390 Chr.12: 52449623-52449623 on Build GRCh38
rs368407469 Chr.12: 52449707-52449707 on Build GRCh38
rs779321597 Chr.12: 52449879-52449879 on Build GRCh38
rs757570546 Chr.12: 52449763-52449763 on Build GRCh38
rs760408900 Chr.12: 52449895-52449895 on Build GRCh38
rs765492618 Chr.12: 52449549-52449549 on Build GRCh38
rs369853484 Chr.12: 52449543-52449543 on Build GRCh38
rs142625176 Chr.12: 52449612-52449612 on Build GRCh38
rs759996259 Chr.12: 52449544-52449544 on Build GRCh38
rs382894 Chr.12: 52449797-52449797 on Build GRCh38
rs774682488 Chr.12: 52449516-52449516 on Build GRCh38
rs151191247 Chr.12: 52449829-52449829 on Build GRCh38
rs772787387 Chr.12: 52449894-52449894 on Build GRCh38
rs780212659 Chr.12: 52449850-52449850 on Build GRCh38
rs552586932 Chr.12: 52449679-52449679 on Build GRCh38
rs774030808 Chr.12: 52449681-52449681 on Build GRCh38
rs745903126 Chr.12: 52449669-52449669 on Build GRCh38
rs753070394 Chr.12: 52449553-52449553 on Build GRCh38
rs778196345 Chr.12: 52449663-52449663 on Build GRCh38
rs765889169 Chr.12: 52449824-52449824 on Build GRCh38
rs533621164 Chr.12: 52449733-52449733 on Build GRCh38
rs572718504 Chr.12: 52449803-52449803 on Build GRCh38
rs772378946 Chr.12: 52449522-52449522 on Build GRCh38
rs373427648 Chr.12: 52449639-52449639 on Build GRCh38
rs756186078 Chr.12: 52449847-52449847 on Build GRCh38
rs771623502 Chr.12: 52449813-52449813 on Build GRCh38
rs374881736 Chr.12: 52449807-52449807 on Build GRCh38
rs749763025 Chr.12: 52449791-52449791 on Build GRCh38
rs763367321 Chr.12: 52449672-52449672 on Build GRCh38
rs749910489 Chr.12: 52449709-52449709 on Build GRCh38
rs145344532 Chr.12: 52449842-52449842 on Build GRCh38
rs758010117 Chr.12: 52449554-52449554 on Build GRCh38
rs368064736 Chr.12: 52449819-52449819 on Build GRCh38
rs145327282 Chr.12: 52449557-52449557 on Build GRCh38
rs762053996 Chr.12: 52449636-52449636 on Build GRCh38
rs997805328 Chr.12: 52449689-52449689 on Build GRCh38
rs753361235 Chr.12: 52449826-52449826 on Build GRCh38
rs182918456 Chr.12: 52449889-52449889 on Build GRCh38
rs551355854 Chr.12: 52449620-52449620 on Build GRCh38
rs747204937 Chr.12: 52449664-52449664 on Build GRCh38
rs779744349 Chr.12: 52449581-52449581 on Build GRCh38
rs370461879 Chr.12: 52449600-52449600 on Build GRCh38
rs370867896 Chr.12: 52449645-52449645 on Build GRCh38
rs776954932 Chr.12: 52449481-52449497 on Build GRCh38
rs377079859 Chr.12: 52449595-52449595 on Build GRCh38
rs766290591 Chr.12: 52449720-52449720 on Build GRCh38
rs673602 Chr.12: 52449882-52449882 on Build GRCh38
rs754812957 Chr.12: 52449651-52449651 on Build GRCh38
rs769165235 Chr.12: 52449878-52449878 on Build GRCh38
rs4079521 Chr.12: 52449523-52449523 on Build GRCh38
rs755228431 Chr.12: 52449472-52449472 on Build GRCh38
rs111359970 Chr.12: 52449755-52449755 on Build GRCh38
rs747149723 Chr.12: 52449586-52449586 on Build GRCh38
rs200181626 Chr.12: 52449649-52449649 on Build GRCh38
rs772910057 Chr.12: 52449818-52449818 on Build GRCh38
rs771115384 Chr.12: 52449594-52449594 on Build GRCh38
rs764558869 Chr.12: 52449782-52449782 on Build GRCh38
rs770443749 Chr.12: 52449897-52449897 on Build GRCh38
rs757970006 Chr.12: 52449482-52449482 on Build GRCh38
rs751075044 Chr.12: 52449561-52449561 on Build GRCh38
rs639374 Chr.12: 52449904-52449904 on Build GRCh38
rs746002443 Chr.12: 52449769-52449769 on Build GRCh38
rs761909527 Chr.12: 52449519-52449519 on Build GRCh38
rs566062730 Chr.12: 52449597-52449597 on Build GRCh38
rs137905665 Chr.12: 52449843-52449843 on Build GRCh38
rs777300878 Chr.12: 52449483-52449483 on Build GRCh38
rs770207095 Chr.12: 52449670-52449670 on Build GRCh38
rs371788244 Chr.12: 52449832-52449832 on Build GRCh38
rs536802368 Chr.12: 52449630-52449630 on Build GRCh38
rs756758177 Chr.12: 52449565-52449565 on Build GRCh38
rs760775348 Chr.12: 52449535-52449535 on Build GRCh38
rs533997961 Chr.12: 52449837-52449838 on Build GRCh38
rs745743945 Chr.12: 52449513-52449513 on Build GRCh38
rs145091946 Chr.12: 52449582-52449582 on Build GRCh38
rs371478761 Chr.12: 52449787-52449787 on Build GRCh38
rs529440101 Chr.12: 52449920-52449920 on Build GRCh38
rs113875880 Chr.12: 52449756-52449756 on Build GRCh38
rs773498316 Chr.12: 52449584-52449584 on Build GRCh38
rs425827 Chr.12: 52449750-52449750 on Build GRCh38
rs140303040 Chr.12: 52449835-52449835 on Build GRCh38
rs767148158 Chr.12: 52449708-52449708 on Build GRCh38
rs763310605 Chr.12: 52449608-52449608 on Build GRCh38
rs900922220 Chr.12: 52449674-52449674 on Build GRCh38
rs1025301155 Chr.12: 52449699-52449699 on Build GRCh38
rs752580660 Chr.12: 52449538-52449538 on Build GRCh38
rs776295408 Chr.12: 52449898-52449898 on Build GRCh38
rs755580131 Chr.12: 52449714-52449714 on Build GRCh38
rs752672659 Chr.12: 52449905-52449905 on Build GRCh38
rs373549008 Chr.12: 52449475-52449475 on Build GRCh38
rs571421123 Chr.12: 52449628-52449628 on Build GRCh38
rs889313201 Chr.12: 52449648-52449648 on Build GRCh38
rs977412515 Chr.12: 52449790-52449790 on Build GRCh38
rs200532105 Chr.12: 52449805-52449805 on Build GRCh38
rs11503946 Chr.12: 52449501-52449501 on Build GRCh38
rs781162204 Chr.12: 52449569-52449569 on Build GRCh38
rs749907517 Chr.12: 52449637-52449637 on Build GRCh38
rs146034933 Chr.12: 52449640-52449640 on Build GRCh38
rs369636090 Chr.12: 52449667-52449667 on Build GRCh38
rs367794230 Chr.12: 52449783-52449783 on Build GRCh38
rs781409851 Chr.12: 52449767-52449767 on Build GRCh38
rs150034493 Chr.12: 52449851-52449851 on Build GRCh38
rs753798928 Chr.12: 52449742-52449742 on Build GRCh38
rs759050587 Chr.12: 52449900-52449900 on Build GRCh38
rs769460857 Chr.12: 52449514-52449514 on Build GRCh38
rs375686236 Chr.12: 52449666-52449666 on Build GRCh38
rs190319305 Chr.12: 52449570-52449570 on Build GRCh38
rs778688221 Chr.12: 52449474-52449474 on Build GRCh38

View Full Product Details