Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10451584-10452124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APITD1-CORT
Entrez Gene ID
100526739
Gene Name
APITD1-CORT readthrough
Gene Aliases
-
UniGene
Hs.412311
Gene Chromosome Location
Chr.1: 10430102-10452003 on Build GRCh38
Transcripts
NM_001243768.1 NM_001270517.1 NM_198544.3 NM_199006.2 NR_037187.1 NM_001302.4

Gene Symbol
CORT
Entrez Gene ID
1325
Gene Name
cortistatin
Gene Aliases
CST-14, CST-17, CST-29
UniGene
Hs.412311
Gene Chromosome Location
Chr.1: 10449719-10452003 on Build GRCh38
Transcripts
NM_001243768.1 NM_001270517.1 NM_198544.3 NM_199006.2 NR_037187.1 NM_001302.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3801106 c.463A>G p.K155E Chr.1: 10451590-10451590 on Build GRCh38
3801105 c.490A>G p.K164E Chr.1: 10451590-10451590 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs554553942 Chr.1: 10451736-10451736 on Build GRCh38
rs76343728 Chr.1: 10451817-10451817 on Build GRCh38
rs893884897 Chr.1: 10452060-10452061 on Build GRCh38
rs755252731 Chr.1: 10451619-10451619 on Build GRCh38
rs757958192 Chr.1: 10451632-10451632 on Build GRCh38
rs1040528950 Chr.1: 10451880-10451880 on Build GRCh38
rs953600186 Chr.1: 10451749-10451749 on Build GRCh38
rs915177063 Chr.1: 10451796-10451796 on Build GRCh38
rs147076752 Chr.1: 10451918-10451918 on Build GRCh38
rs924737112 Chr.1: 10452097-10452097 on Build GRCh38
rs766732752 Chr.1: 10451617-10451617 on Build GRCh38
rs946390415 Chr.1: 10451813-10451813 on Build GRCh38
rs375360298 Chr.1: 10451611-10451611 on Build GRCh38
rs1009179110 Chr.1: 10451772-10451773 on Build GRCh38
rs750998110 Chr.1: 10452028-10452028 on Build GRCh38
rs1027897624 Chr.1: 10451959-10451959 on Build GRCh38
rs1019226082 Chr.1: 10451797-10451797 on Build GRCh38
rs1029444378 Chr.1: 10451744-10451744 on Build GRCh38
rs766307113 Chr.1: 10451699-10451702 on Build GRCh38
rs138494067 Chr.1: 10451979-10451979 on Build GRCh38
rs893884522 Chr.1: 10451729-10451729 on Build GRCh38
rs552286959 Chr.1: 10452085-10452085 on Build GRCh38
rs888845491 Chr.1: 10451840-10451840 on Build GRCh38
rs541272987 Chr.1: 10451845-10451845 on Build GRCh38
rs570483788 Chr.1: 10452096-10452096 on Build GRCh38
rs192809834 Chr.1: 10451618-10451618 on Build GRCh38
rs962175602 Chr.1: 10451816-10451816 on Build GRCh38
rs778587456 Chr.1: 10451620-10451620 on Build GRCh38
rs540895114 Chr.1: 10451948-10451948 on Build GRCh38
rs1307497 Chr.1: 10451999-10451999 on Build GRCh38
rs1045523470 Chr.1: 10451621-10451621 on Build GRCh38
rs530566578 Chr.1: 10452029-10452029 on Build GRCh38
rs915601626 Chr.1: 10451921-10451925 on Build GRCh38
rs1054736808 Chr.1: 10452048-10452048 on Build GRCh38
rs949736695 Chr.1: 10451978-10451978 on Build GRCh38
rs952424250 Chr.1: 10451970-10451970 on Build GRCh38
rs752339274 Chr.1: 10451624-10451624 on Build GRCh38
rs999106415 Chr.1: 10451698-10451698 on Build GRCh38
rs560492454 Chr.1: 10451863-10451863 on Build GRCh38
rs1052301359 Chr.1: 10451776-10451776 on Build GRCh38
rs374282943 Chr.1: 10452009-10452009 on Build GRCh38
rs377518871 Chr.1: 10451865-10451865 on Build GRCh38
rs971472464 Chr.1: 10452019-10452019 on Build GRCh38
rs981881639 Chr.1: 10451737-10451737 on Build GRCh38
rs897994438 Chr.1: 10451900-10451900 on Build GRCh38
rs529960906 Chr.1: 10451957-10451957 on Build GRCh38
rs558667685 Chr.1: 10451879-10451879 on Build GRCh38
rs184922802 Chr.1: 10451732-10451732 on Build GRCh38
rs927809674 Chr.1: 10451768-10451768 on Build GRCh38
rs935211593 Chr.1: 10451771-10451771 on Build GRCh38
rs924537181 Chr.1: 10451988-10451988 on Build GRCh38
rs76990442 Chr.1: 10451773-10451773 on Build GRCh38
rs559569899 Chr.1: 10451955-10451955 on Build GRCh38
rs751370893 Chr.1: 10451701-10451702 on Build GRCh38
rs951920815 Chr.1: 10451906-10451906 on Build GRCh38

View Full Product Details