Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 20187351-20187887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJB2
Entrez Gene ID
2706
Gene Name
gap junction protein beta 2
Gene Aliases
CX26, DFNA3, DFNA3A, DFNB1, DFNB1A, HID, KID, NSRD1, PPK
UniGene
Hs.524894
Gene Chromosome Location
Chr.13: 20187463-20192975 on Build GRCh38
Transcripts
NM_004004.5 XM_011535049.2

SNP Detail

SNP ID Chromosome Location
rs1000548264 Chr.13: 20187422-20187422 on Build GRCh38
rs563627004 Chr.13: 20187420-20187420 on Build GRCh38
rs11839674 Chr.13: 20187454-20187454 on Build GRCh38
rs1016745684 Chr.13: 20187662-20187662 on Build GRCh38
rs886050025 Chr.13: 20187607-20187610 on Build GRCh38
rs1027416752 Chr.13: 20187765-20187765 on Build GRCh38
rs1054537176 Chr.13: 20187557-20187557 on Build GRCh38
rs966209198 Chr.13: 20187569-20187569 on Build GRCh38
rs562612904 Chr.13: 20187594-20187594 on Build GRCh38
rs1003447976 Chr.13: 20187683-20187683 on Build GRCh38
rs959525463 Chr.13: 20187692-20187692 on Build GRCh38
rs944937219 Chr.13: 20187413-20187413 on Build GRCh38
rs9237 Chr.13: 20187834-20187834 on Build GRCh38
rs891851629 Chr.13: 20187559-20187559 on Build GRCh38
rs7988691 Chr.13: 20187624-20187624 on Build GRCh38
rs1042285772 Chr.13: 20187400-20187400 on Build GRCh38
rs995031937 Chr.13: 20187753-20187753 on Build GRCh38
rs1039460729 Chr.13: 20187417-20187420 on Build GRCh38
rs751581043 Chr.13: 20187643-20187643 on Build GRCh38
rs7623 Chr.13: 20187749-20187749 on Build GRCh38
rs752926267 Chr.13: 20187819-20187819 on Build GRCh38
rs917708342 Chr.13: 20187741-20187741 on Build GRCh38
rs531576198 Chr.13: 20187436-20187436 on Build GRCh38
rs180788420 Chr.13: 20187532-20187532 on Build GRCh38
rs1007729682 Chr.13: 20187515-20187515 on Build GRCh38
rs886050026 Chr.13: 20187695-20187695 on Build GRCh38
rs1053932829 Chr.13: 20187441-20187441 on Build GRCh38
rs912033122 Chr.13: 20187861-20187861 on Build GRCh38
rs902152201 Chr.13: 20187408-20187408 on Build GRCh38
rs921942929 Chr.13: 20187452-20187452 on Build GRCh38
rs949269226 Chr.13: 20187379-20187379 on Build GRCh38
rs950749711 Chr.13: 20187631-20187631 on Build GRCh38
rs11841182 Chr.13: 20187704-20187704 on Build GRCh38
rs535822487 Chr.13: 20187814-20187814 on Build GRCh38
rs977947783 Chr.13: 20187769-20187769 on Build GRCh38
rs1017820745 Chr.13: 20187519-20187519 on Build GRCh38

View Full Product Details